Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ LRP1, Rabbit, Unlabeled, Polyclonal,

Catalog No. ABN1018MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABN1018MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN1018MI Supplier MilliporeSigma™ Supplier No. ABN1018
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Prolow-density lipoprotein receptor-related protein 1 (UniProt Q07954; also known as A2MR, Alpha-2-macroglobulin receptor, APOER, Apolipoprotein E receptor, CD91, LRP-1) is encoded by the LRP1 (also known as A2MR, APR) gene (Gene ID 4035) in human. LRP1 is an LDLR family member expressed by vascular endothelial cells forming the blood-brain barrier (BBB), vascular mural cells (pericytes and vascular smooth muscle cells (VSMCs)), neurons, and astrocytes. LRP1 recognizes and mediates the endocytosis of more than 40 different ligands, including apolipoprotein E (apoE), APP and Aβ. LRP1 is the primary receptor mediating transport of Aβ across the BBB into circulation, thereby clearing it from the brain. LRP1 is synthesized with a signal peptide sequence (a.a. 1-19), posttranslational cleavage yields a soluble form (sLRP1) and a cell surface-bound form composed of an 515kDa extracellular α-chain (LRP-515) and a 85kDa single-transmembrane (a.a. 4420-4444) β-chain (LRP-85) that contains the 100-amino intracellular domain (LRPICD; a.a. 4445-4544). BACE-1, ADAM-10, ADAM-12, ADAM-17, MT1-MMP, and t-PA are shown to be involved in the shedding of LRP1 to produce sLRP1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRP1
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Rabbit polyclonal antibody serum with 0.05% Sodium Azide.
Gene LRP1, A2MR, APR
Gene Accession No. Q07954
Host Species Rabbit
Immunogen GST-tagged recombinant human LRP1 intracellular domain (LRPICD).
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_002323
Test Specificity Recognizes the Pro- and mature LRP-1 (a.a. 1-4544 and 20-4544, respectively), the beta-secretase-cleaved, membrane-bound LRP-85, and the gamma-secretase-cleaved intracellular domain LRPICD fragment, but not the beta-secretase-cleaved extracellular LRP-515 fragment.
Target Species Mouse
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Serum
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.