Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRP10 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300097AF488
Description
LRP10 Polyclonal specifically detects LRP10 in Human samples. It is validated for ELISA.Specifications
LRP10 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
26020 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
Alexa Fluor 488 | |
DKFZp564C1940, low density lipoprotein receptor-related protein 10, low-density lipoprotein receptor-related protein 10, LRP-10, LRP9, MGC142274, MGC142276, MGC8675, MST087 | |
Produced in rabbits immunized with purified, recombinant LRP10 (Q7Z4F1-1; Met1-Lys440). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction