Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRP10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581908
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581908 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581908 Supplier Invitrogen™ Supplier No. PA581908
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PNHACEDPPA VLLEVQGTLQ RPLVRDSRTS PANCTWLILG SKEQTVTIRF QKLHLACGSE RLTLRSPLQP LISLCEAPPS PLQLPGGNVT ITYSYAGARA PMGQGFLLSY SQDWLMCLQE EFQCLNHRCV SAVQRCDG Highest antigen sequence indentity to the following orthologs: Mouse - 86%, Rat - 86%.

Probable receptor, which is involved in the internalization of lipophilic molecules and/or signal transduction. May be involved in the uptake of lipoprotein APOE in liver.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRP10
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LRP10
Gene Accession No. Q7Z4F1
Gene Alias LDL receptor related protein 10; low-density lipoprotein receptor-related protein 10; LRP10; LRP-10; Lrp9; MST087; MSTP087; SP220; UNQ389/PRO724
Gene Symbols LRP10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LRP10. Recombinant protein control fragment (Product #RP-89007).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26020
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.