Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRP12 Monoclonal Antibody (4E11C5)
GREENER_CHOICE

Catalog No. PIMA538561
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538561 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538561 Supplier Thermo Scientific Supplier No. MA538561
Add to Cart
Add to Cart

LRP12 Monoclonal Antibody (4E11C5)

LRP12 Monoclonal Antibody for Western Blot, Flow, ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen LRP12
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 4E11C5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene LRP12
Gene Accession No. Q9Y561
Gene Alias AI848829; C820005L12Rik; LDL receptor related protein 12; LDLR-related protein 12; low density lipoprotein receptor-related protein 12; low density lipoprotein-related protein 12; low-density lipoprotein receptor-related protein 12; LRP12; LRP-12; ST7; Suppressor of tumorigenicity 7 protein
Gene Symbols LRP12
Host Species Mouse
Immunogen Purified recombinant fragment of human LRP12 (aa: extra 355-492) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29967
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.