Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LRP12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550233
Description
The antibody detects endogenous levels of total LRP12 protein.
Members of the LDL receptor gene family, including LDLR (low density lipoprotein receptor), LRP1 (low density lipoprotein related protein), Megalin (also designated GP330), VLDLR (very low density lipoprotein receptor) and ApoER2 are characterized by a cluster of cysteine-rich class A repeats, epidermal growth factor (EGF)-like repeats, YWTD repeats and an O-linked sugar domain. LRP12, also designated ST7, is a member of the LDLR family that is thought to be involved in the internalization of lipophilic molecules and/or signal transduction. LRP12 has been shown to interact with RACK1, MADHIP and Integrin beta-1-binding protein 3. It is widely expressed in heart, skeletal muscle, brain, lung, placenta and pancreas. Overexpression of LRP12 may be associated with oral tumors, therefore implicating LRP12 as a potential oncogene.
Specifications
| LRP12 | |
| Polyclonal | |
| Unconjugated | |
| LRP12 | |
| AI848829; C820005L12Rik; LDL receptor related protein 12; LDLR-related protein 12; low density lipoprotein receptor-related protein 12; low density lipoprotein-related protein 12; low-density lipoprotein receptor-related protein 12; LRP12; LRP-12; ST7; Suppressor of tumorigenicity 7 protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 29967 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.8 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9Y561 | |
| LRP12 | |
| Fusion protein corresponding to a region derived from internal residues of human low density lipoprotein receptor-related protein 12. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction