Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRP130 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988716
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8716 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8716 Supplier Bioss Supplier No. BS8719R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a deadenylase that functions as the catalytic subunit of the polyadenylate binding protein dependent poly(A) nuclease complex. The encoded protein is a magnesium dependent 3′ to 5′ exoribonuclease that is involved in the degradation of cytoplasmic mRNAs. Alternate splicing results in multiple transcript variants.

Specifications

Antigen LRP130
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene LRPPRC
Gene Accession No. P42704
Gene Alias 130 kDa leucine-rich protein; 3110001K13Rik; C76645; CLONE-23970; GP130; leucine rich pentatricopeptide repeat containing; Leucine rich protein 157; leucine rich protein LRP130; leucine-rich pentatricopeptide repeat containing; Leucine-rich PPR motif-containing protein, mitochondrial; leucine-rich PPR-motif containing; leucine-rich protein 157; LRP 130; Lrp130; Lrp157; LRPPRC; LSFC; mitochondrial leucine-rich PPR motif-containing protein; rLRP157
Gene Symbols LRPPRC
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human LRP130.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10128
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.