Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRP1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547836
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547836 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547836 Supplier Invitrogen™ Supplier No. PA547836
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) LRP-1, rhLRP-4, rhLRP-5, and rhLRP-6 is observed. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.

This gene encodes a member of the low density lipoprotein (LDL) receptor family. These receptors play a wide variety of roles in normal cell function and development due to their interactions with multiple ligands. Disruption of this gene has been reported in several types of cancer. [provided by RefSeq, Jun 2016].
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRP1B
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene LRP1B
Gene Accession No. Q9NZR2
Gene Alias 9630004P12Rik; EC 1.1.1.94; EC 3.4.21.9; LDL receptor related protein 1B; low density lipoprotein receptor related protein-deleted in tumor; low density lipoprotein receptor-related protein 1B; low density lipoprotein-related protein 1B (deleted in tumors); low-density lipoprotein receptor-related protein 1B; Low-density lipoprotein receptor-related protein-deleted in tumor; LRP1B; LRP-1B; LRP-deleted in tumors; LRPDIT; LRP-DIT
Gene Symbols LRP1B
Host Species Sheep
Immunogen E. coli-derived recombinant human LRP-1B Leu21-Glu244.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53353
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.