Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRP2 Antibody (545818), FITC, R&D Systems™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals MAB95781F
Description
LRP2 Monoclonal specifically detects LRP2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
LRP2 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
LRP2 | |
Mouse myeloma cell line NS0-derived partial recombinant human Megalin/LRP2 protein (amino acids 3510-3964) | |
0.1 mL | |
Lipid and Metabolism | |
Human | |
IgG1 |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
545818 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
calcium sensor protein, DBSlow-density lipoprotein receptor-related protein 2, EC 1.1.2.3, EC 3.4.21.9, Glycoprotein 330, GP330, Heymann nephritis antigen homolog, low density lipoprotein receptor-related protein 2, LRP-2, megalin | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction