Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564182
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564182 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564182 Supplier Invitrogen™ Supplier No. PA564182
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NNDRIYWSDF KEDVIETIKY DGTDRRVIAK EAMNPYSLDI FEDQLYWISK EKGEVWKQNK FGQGKKEKTL VVNPWLTQVR IFHQLRYNKS VP Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 79%.

The protein encoded by this gene, low density lipoprotein-related protein 2 (LRP2) or megalin, is a multi-ligand endocytic receptor that is expressed in many different tissues but primarily in absorptive epithilial tissues such as the kidney. This glycoprotein has a large amino-terminal extracellular domain, a single transmembrane domain, and a short carboxy-terminal cytoplasmic tail. The extracellular ligand-binding-domains bind diverse macromolecules including albumin, apolipoproteins B and E, and lipoprotein lipase. The LRP2 protein is critical for the reuptake of numerous ligands, including lipoproteins, sterols, vitamin-binding proteins, and hormones. This protein also has a role in cell-signaling; extracellular ligands include parathyroid horomones and the morphogen sonic hedgehog while cytosolic ligands include MAP kinase scaffold proteins and JNK interacting proteins. Recycling of this membrane receptor is regulated by phosphorylation of its cytoplasmic domain. Mutations in this gene cause Donnai-Barrow syndrome (DBS) and facio-oculoacoustico-renal syndrome (FOAR).
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRP2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Lrp2
Gene Accession No. P98164
Gene Alias AI315343; AW536255; b2b1625.2Clo; calcium sensor protein; D230004K18Rik; DBS; Glycoprotein 330; Gp330; Heymann nephritis antigen homolog; LDL receptor related protein 2; low density lipoprotein receptor-related protein 2; low density lipoprotein-related protein 2; low-density lipoprotein receptor-related protein 2; Lrp2; LRP-2; Megalin
Gene Symbols Lrp2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LRP2. Recombinant protein control fragment (Product #RP-103570).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4036
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.