Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LRP2 Recombinant Rabbit Monoclonal Antibody (10B5)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA550033
Description
LRP2 Recombinant Monoclonal Antibody for Flow, ELISA
The protein encoded by this gene, low density lipoprotein-related protein 2 (LRP2) or megalin, is a multi-ligand endocytic receptor that is expressed in many different tissues but primarily in absorptive epithilial tissues such as the kidney. This glycoprotein has a large amino-terminal extracellular domain, a single transmembrane domain, and a short carboxy-terminal cytoplasmic tail. The extracellular ligand-binding-domains bind diverse macromolecules including albumin, apolipoproteins B and E, and lipoprotein lipase. The LRP2 protein is critical for the reuptake of numerous ligands, including lipoproteins, sterols, vitamin-binding proteins, and hormones. This protein also has a role in cell-signaling; extracellular ligands include parathyroid horomones and the morphogen sonic hedgehog while cytosolic ligands include MAP kinase scaffold proteins and JNK interacting proteins. Recycling of this membrane receptor is regulated by phosphorylation of its cytoplasmic domain. Mutations in this gene cause Donnai-Barrow syndrome (DBS) and facio-oculoacoustico-renal syndrome (FOAR).
Specifications
LRP2 | |
Recombinant Monoclonal | |
3 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P98164 | |
Lrp2 | |
A synthesized peptide derived from human LRP2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Flow Cytometry | |
10B5 | |
Unconjugated | |
Lrp2 | |
AI315343; AW536255; b2b1625.2Clo; calcium sensor protein; D230004K18Rik; DBS; Glycoprotein 330; Gp330; Heymann nephritis antigen homolog; LDL receptor related protein 2; low density lipoprotein receptor-related protein 2; low density lipoprotein-related protein 2; low-density lipoprotein receptor-related protein 2; Lrp2; LRP-2; Megalin | |
Rabbit | |
Affinity chromatography | |
RUO | |
4036 | |
-20°C or -80°C if preferred | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction