Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRP2BP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569181
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569181 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569181 Supplier Invitrogen™ Supplier No. PA569181
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%.

May act as an adapter that regulates LRP2 function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRP2BP
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LRP2BP
Gene Accession No. Q569C2, Q9P2M1
Gene Alias 1700113N17Rik; 2310006J04Rik; 4930479L12Rik; KIAA1325; low density lipoprotein receptor-related protein 2 binding protein; Lrp2 binding protein; LRP2-binding protein; Lrp2bp; Megalin-binding protein; megBP; RGD1305896; zgc:165631
Gene Symbols LRP2BP
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human LRP2BP (aa 201-250).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 290753, 55805
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.