Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA536486
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536486 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536486 Supplier Invitrogen™ Supplier No. PA536486
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 80 kDa. Purity is >95% by SDS-PAGE.

LRP3 is a probable receptor, widely expressed, which may be involved in the internalization of lipophilic molecules and/or signal transduction. Its precise role is, however unclear, since it does not bind to very low density lipoprotein (VLDL) or to LRPAP1 in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRP3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene LRP3
Gene Accession No. O75074, O88204
Gene Alias 105 kDa low-density lipoprotein receptor-related protein; CTD-2540B15.13; hLRp105; LDL receptor related protein 3; low density lipoprotein receptor-related protein 3; low-density lipoprotein receptor-related protein 3; Lrp105; LRP3; LRP-3; rLRp105
Gene Symbols LRP3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 31-80 of human LRP3.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4037, 435965, 89787
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.