Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRP3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA539230
Description
LRP3 is a probable receptor, widely expressed, which may be involved in the internalization of lipophilic molecules and/or signal transduction. Its precise role is, however unclear, since it does not bind to very low density lipoprotein (VLDL) or to LRPAP1 in vitro.
Specifications
| LRP3 | |
| Polyclonal | |
| Unconjugated | |
| LRP3 | |
| Low-density lipoprotein receptor-related protein 3, LRP-3, 105 kDa low-density lipoprotein receptor-related protein, hLRp105, LRP3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4037, 435965, 89787 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75074, O88204 | |
| LRP3 | |
| A synthetic peptide derived from the internal region of human LRP3 | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction