Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRP4 Monoclonal Antibody (N207/27)
GREENER_CHOICE

Catalog No. PIMA527675
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527675 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527675 Supplier Invitrogen™ Supplier No. MA527675
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

1 μg/mL of MA5-27675 was sufficient for detection of LRP4 in 20 μg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects the extracellular domain of LRP4 approximately 215kDa. Degradation product can be seen at approximately 150kDa. This antibody was formerly sold as clone S207-27.

This gene encodes a member of the low-density lipoprotein receptor-related protein family. The encoded protein may be a regulator of Wnt signaling. Mutations in this gene are associated with Cenani-Lenz syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRP4
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone N207/27
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
Gene LRP4
Gene Accession No. O75096, Q8VI56, Q9QYP1
Gene Alias 6430526J12Rik; CLSS; CMS17; Corin; D230026E03; Kiaa0816; LDL receptor related protein 4; LDLR; LDLR dan; low density lipoprotein receptor-related protein 4; low-density lipoprotein receptor-related protein 4; LRP10; Lrp4; LRP-4; mdig; Megf7; multiple epidermal growth factor-like domains 7; SOST2
Gene Symbols LRP4
Host Species Mouse
Immunogen Fusion protein amino acids 26- 350 (extracellular N-terminus) of mouse LTP4.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 228357, 4038, 83469
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.