Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568218
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568218 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568218 Supplier Invitrogen™ Supplier No. PA568218
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Human and Rat samples.

This gene encodes a member of the low-density lipoprotein receptor-related protein family. The encoded protein may be a regulator of Wnt signaling. Mutations in this gene are associated with Cenani-Lenz syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRP4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 30% glycerol, 0.87% NaCl and 0.01% sodium azide; pH 7.3
Gene LRP4
Gene Accession No. O75096, Q8VI56, Q9QYP1
Gene Alias 6430526J12Rik; CLSS; CMS17; Corin; D230026E03; Kiaa0816; LDL receptor related protein 4; LDLR; LDLR dan; low density lipoprotein receptor-related protein 4; low-density lipoprotein receptor-related protein 4; LRP10; Lrp4; LRP-4; mdig; Megf7; multiple epidermal growth factor-like domains 7; SOST2
Gene Symbols LRP4
Host Species Rabbit
Immunogen Synthesized peptide derived from human LRP4.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 228357, 4038, 83469
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.