Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118912
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118912 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5118912 Supplier Invitrogen™ Supplier No. PA5118912
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total LRP4.

This gene encodes a member of the low-density lipoprotein receptor-related protein family. The encoded protein may be a regulator of Wnt signaling. Mutations in this gene are associated with Cenani-Lenz syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRP4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene LRP4
Gene Accession No. O75096, Q8VI56, Q9QYP1
Gene Alias 6430526J12Rik; CLSS; CMS17; Corin; D230026E03; Kiaa0816; LDL receptor related protein 4; LDLR; LDLR dan; low density lipoprotein receptor-related protein 4; low-density lipoprotein receptor-related protein 4; LRP10; Lrp4; LRP-4; mdig; Megf7; multiple epidermal growth factor-like domains 7; SOST2
Gene Symbols LRP4
Host Species Rabbit
Immunogen A synthesized peptide derived from human LRP4(Accession O75096), corresponding to amino acid residues T271-N321.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 228357, 4038, 83469
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.