Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRP5 Mouse anti-Human, Clone:2B11, Abnova™

Catalog No. 89334569 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-569 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-569 Supplier Abnova Corporation Supplier No. MAB17714
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human LRP5.

Specifications

Antigen LRP5
Applications ELISA, Flow Cytometry
Classification Monoclonal
Clone 2B11
Conjugate Unconjugated
Description low density lipoprotein receptor-related protein 5
Dilution ELISA (1:10000)
Flow Cytometry (1:200-1:400)
Immunocytochemistry
Immunohistochemistry
Western Blot (1:500-1:2000)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene LRP5
Gene Alias BMND1, EVR1, EVR4, HBM, LR3, LRP7, OPPG, OPS, OPTA1, VBCH2
Host Species Mouse
Immunogen Recombinant protein corresponding to human LRP5 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 4041
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.