Learn More
Invitrogen™ LRP5 Polyclonal Antibody

Description
LRP5 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | LRP5 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | LRP5 |
| Gene Accession No. | O75197, Q91VN0 |
| Gene Alias | BMND1; EVR1; EVR4; HBM; HGNC:6697; LDL receptor related protein 5; low density lipoprotein receptor-related protein 5; low density lipoprotein receptor-related protein 7; low-density lipoprotein receptor-related protein 5; Low-density lipoprotein receptor-related protein 7; LR3; Lrp5; LRP-5; LRP7; LRP-7; mKIAA4142; OPPG; OPS; OPTA1; VBCH2 |
| Gene Symbols | LRP5 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.