Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRP5L Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981498
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1498 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1498 Supplier Bioss Supplier No. BS11590R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen LRP5L
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene LRP5L
Gene Accession No. A4QPB2
Gene Alias LDL receptor related protein 5 like; low density lipoprotein receptor-related protein 5-like; Low-density lipoprotein receptor-related protein 5-like protein; LRP5L; LRP-5-like
Gene Symbols LRP5L
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human LRP5L.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91355
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.