Learn More
Invitrogen™ LRP6 Monoclonal Antibody (1A3F5)

Description
This antibody has been tested in indirect ELISA.
Specifications
Specifications
| Antigen | LRP6 |
| Applications | ELISA, Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | 1A3F5 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% sodium azide |
| Gene | LRP6 |
| Gene Accession No. | O75581 |
| Gene Alias | ADCAD2; C030016K15Rik; Cd; crooked; FLJ90062; FLJ90421; Gw; LDL receptor related protein 6; low density lipoprotein receptor-related protein 6; low density lipoprotein receptor-related protein 6; low density lipoprotein-related protein 6; low density lipoprotein-related protein 6; low-density lipoprotein receptor-related protein 6; lpr6; LRP6; LRP-6; ska>m26Jus>; ska26; skax26; STHAG7 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.