Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LRP6 Monoclonal Antibody (290913)
Rat Monoclonal Antibody
Supplier: Invitrogen™ MA523995
Description
In direct ELISAs, this antibody shows approximately 50-60% cross-reactivity with recombinant human (rh) LRP-6 and no cross-reactivity with rhLDL R. Reconstitute at 0.5 mg/mL in sterile PBS.
LRP6 is essential for the Wnt/beta catenin signaling pathway, probably by acting as a coreceptor together with Frizzled for Wnt. It is a specific, high-affinity receptor for DKK1 and DKK2, but not DKK3. The interaction with DKK1 blocks LRP6-mediated Wnt/beta catenin signaling via LRP6 removal via Kremen proteins-mediated endocytosis.
Specifications
LRP6 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
O88572 | |
LRP6 | |
CHO-derived recombinant mouse LRP-6 Met1-Pro1368. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2b |
Western Blot | |
290913 | |
Unconjugated | |
LRP6 | |
ADCAD2; C030016K15Rik; Cd; crooked; FLJ90062; FLJ90421; Gw; LDL receptor related protein 6; low density lipoprotein receptor-related protein 6; low density lipoprotein receptor-related protein 6; low density lipoprotein-related protein 6; low density lipoprotein-related protein 6; low-density lipoprotein receptor-related protein 6; lpr6; LRP6; LRP-6; ska>m26Jus>; ska26; skax26; STHAG7 | |
Rat | |
Protein A/G | |
RUO | |
16974 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction