Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRP6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589161
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589161 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589161 Supplier Invitrogen™ Supplier No. PA589161
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: APLLLYANRR DLRLVDATNG KENATIVVGG LEDAAAVDFV FSHGLIYWSD VSEEAIKRTE FNKTESVQNV VVSGLLSPDG LACDWLGEKL YWTDSETNRI EVSNLDGSLR KVLFWQELDQ PRAIALDPSS G; Positive Samples: Mouse liver; Cellular Location: Endoplasmic reticulum, Membrane, Single-pass type I membrane protein.

LRP6 is essential for the Wnt/beta catenin signaling pathway, probably by acting as a coreceptor together with Frizzled for Wnt. It is a specific, high-affinity receptor for DKK1 and DKK2, but not DKK3. The interaction with DKK1 blocks LRP6-mediated Wnt/beta catenin signaling via LRP6 removal via Kremen proteins-mediated endocytosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRP6
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.14 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene LRP6
Gene Accession No. O75581, O88572
Gene Alias ADCAD2; C030016K15Rik; Cd; crooked; FLJ90062; FLJ90421; Gw; LDL receptor related protein 6; low density lipoprotein receptor-related protein 6; low density lipoprotein receptor-related protein 6; low density lipoprotein-related protein 6; low density lipoprotein-related protein 6; low-density lipoprotein receptor-related protein 6; lpr6; LRP6; LRP-6; ska>m26Jus>; ska26; skax26; STHAG7
Gene Symbols LRP6
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 20-150 of human LRP6 (NP_0023272).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16974, 4040
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.