Learn More
Invitrogen™ LRP8 Polyclonal Antibody

Description
Immunogen sequence: PLWAEPCLGE TREPEDPAPA LKELFVLPGE PRSQLHQLPK NPLSELPVVK SKRVALSLED DGLP; Positive Samples: SH-SY5Y, Rat testis; Cellular Location: Cell membrane, Single-pass type I membrane protein.
Specifications
Specifications
| Antigen | LRP8 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.58 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | LRP8 |
| Gene Accession No. | Q14114 |
| Gene Alias | 4932703M08Rik; AA921429; AI848122; ApoE receptor 2; APOER2; Apolipoprotein E receptor 2; HSZ75190; LDL receptor related protein 8; low density lipoprotein receptor relative, 8 ligand binding repeats, brain; low density lipoprotein receptor-related protein 8, apolipoprotein e receptor; low-density lipoprotein receptor-related protein 8; Lr8b; Lrp8; LRP-8; MCI1; Protein LR8B |
| Gene Symbols | LRP8 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.