Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRP8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598455
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598455 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598455 Supplier Invitrogen™ Supplier No. PA598455
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LRP8 Polyclonal Antibody for ELISA, ICC/IF, Western Blot

Apolipoprotein E receptor 2 (ApoER2) is an NMDA associated protein found in neurons. It participates in important functions from guiding neurons in the embryonic brain to regulating synaptic signaling. It is also involved in debilitating neurodegenerative diseases such as schizophrenia and Alzheimer's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRP8
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene LRP8
Gene Accession No. Q14114
Gene Alias 4932703M08Rik; AA921429; AI848122; ApoE receptor 2; APOER2; Apolipoprotein E receptor 2; HSZ75190; LDL receptor related protein 8; low density lipoprotein receptor relative, 8 ligand binding repeats, brain; low density lipoprotein receptor-related protein 8, apolipoprotein e receptor; low-density lipoprotein receptor-related protein 8; Lr8b; Lrp8; LRP-8; MCI1; Protein LR8B
Gene Symbols LRP8
Host Species Rabbit
Immunogen Recombinant Human Low-density lipoprotein receptor-related protein 8 protein (131-332AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7804
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.