Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRP8 Recombinant Rabbit Monoclonal Antibody (0V1R8)
GREENER_CHOICE

Catalog No. PIMA538208
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538208 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538208 Supplier Invitrogen™ Supplier No. MA538208
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: U-87MG, Mouse testis, Rat testis Immunogen sequence: PVYRKTTEEE DEDELHIGRT AQIGHVYPAA ISSFDRPLWA EPCLGETREP EDPAPALKEL FVLPGEPRSQ LHQLPKNPLS ELPVVKSKRV ALSLEDDGLP.

Apolipoprotein E receptor 2 (ApoER2) is an NMDA associated protein found in neurons. It participates in important functions from guiding neurons in the embryonic brain to regulating synaptic signaling. It is also involved in debilitating neurodegenerative diseases such as schizophrenia and Alzheimer's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRP8
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 0V1R8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.3
Gene LRP8
Gene Accession No. Q14114, Q924X6
Gene Alias 4932703M08Rik; AA921429; AI848122; ApoE receptor 2; APOER2; Apolipoprotein E receptor 2; HSZ75190; LDL receptor related protein 8; low density lipoprotein receptor relative, 8 ligand binding repeats, brain; low density lipoprotein receptor-related protein 8, apolipoprotein e receptor; low-density lipoprotein receptor-related protein 8; Lr8b; Lrp8; LRP-8; MCI1; Protein LR8B
Gene Symbols LRP8
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16975, 362558, 7804
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.