Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRPAP Antibody (7F1), FITC, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP312047F
Description
LRPAP Monoclonal antibody specifically detects LRPAP in Human samples. It is validated for ELISA, Immunohistochemistry, ImmunofluorescenceSpecifications
LRPAP | |
Monoclonal | |
FITC | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
4043 | |
Store at 4°C in the dark. |
ELISA, Immunohistochemistry, Immunofluorescence | |
7F1 | |
ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
A2MRAPMRAP, A2RAP, alpha-2-macroglobulin receptor-associated protein, alpha-2-macroglobulin receptor-associated protein 1, Alpha-2-MRAP, HBP44, lipoprotein receptor associated protein, low density lipoprotein receptor-related protein associated protein 1, Low density lipoprotein receptor-related protein-associated protein 1, low density lipoprotein-related protein-associated protein 1(alpha-2-macroglobulin receptor-associated protein 1), MGC138272, RAP | |
This antibody was raised by immunising mice with human placental LRPAP. | |
0.1 mL | |
Primary | |
Human | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction