Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRPAP Antibody (8G8), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242682APCCY7
Description
LRPAP Monoclonal specifically detects LRPAP in Human samples. It is validated for Flow Cytometry.Specifications
LRPAP | |
Monoclonal | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
LRPAP1 | |
Recombinant human LRPAP1 (35-357aa) purified from E. coli | |
0.1 mL | |
4043.0 | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
8G8 | |
Flow Cytometry | |
A2MRAPMRAP, A2RAP, alpha-2-macroglobulin receptor-associated protein, alpha-2-macroglobulin receptor-associated protein 1, Alpha-2-MRAP, HBP44, lipoprotein receptor associated protein, low density lipoprotein receptor-related protein associated protein 1, Low density lipoprotein receptor-related protein-associated protein 1, low density lipoprotein-related protein-associated protein 1(alpha-2-macroglobulin receptor-associated protein 1), MGC138272, RAP | |
Mouse | |
Protein A purified | |
Primary | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction