Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRPAP Mouse anti-Human, Clone: 7F1, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP3120460.2MG
Description
LRPAP Monoclonal antibody specifically detects LRPAP in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, ImmunoprecipitationSpecifications
LRPAP | |
Monoclonal | |
1 mg/mL | |
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation | |
A2MRAPMRAP, A2RAP, alpha-2-macroglobulin receptor-associated protein, alpha-2-macroglobulin receptor-associated protein 1, Alpha-2-MRAP, HBP44, lipoprotein receptor associated protein, low density lipoprotein receptor-related protein associated protein 1, Low density lipoprotein receptor-related protein-associated protein 1, low density lipoprotein-related protein-associated protein 1(alpha-2-macroglobulin receptor-associated protein 1), MGC138272, RAP | |
This antibody was raised by immunising mice with human placental LRPAP. | |
0.2 mg | |
Primary | |
Human | |
IgG1 κ |
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation | |
7F1 | |
Unconjugated | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
4043 | |
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction