Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRPAP Rabbit anti-Human, Clone: 4, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
Antigen | LRPAP |
---|---|
Clone | 4 |
Dilution | Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunoprecipitation 1-4 ul/mg of lysate |
Applications | Western Blot, ELISA, Immunofluorescence, Immunoprecipitation |
Classification | Monoclonal |
Description
LRPAP Monoclonal specifically detects LRPAP in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
LRPAP | |
Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunoprecipitation 1-4 ul/mg of lysate | |
Monoclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
4043 | |
IgG | |
Protein A purified |
4 | |
Western Blot, ELISA, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human | |
A2MRAPMRAP, A2RAP, alpha-2-macroglobulin receptor-associated protein, alpha-2-macroglobulin receptor-associated protein 1, Alpha-2-MRAP, HBP44, lipoprotein receptor associated protein, low density lipoprotein receptor-related protein associated protein 1, Low density lipoprotein receptor-related protein-associated protein 1, low density lipoprotein-related protein-associated protein 1(alpha-2-macroglobulin receptor-associated protein 1), MGC138272, RAP | |
Recombinant Human LRPAP protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title