Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567045
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567045 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567045 Supplier Invitrogen™ Supplier No. PA567045
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LDDNELIQFPC KIGQLINLRF LSAARNKLPF LPSEFRNLSL EYLDLFGNTF EQPKVLPVIK LQAPLTLLES SART Highest antigen sequence identity to the following orthologs - mouse 76%, rat 72%.

May negatively regulate the 4-1BB-mediated signaling cascades which result in the activation of NK-kappaB and JNK1. Probable substrate recognition subunit of an ECS (Elongin BC- CUL2/5-SOCS-box protein) E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRR1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LRR1
Gene Accession No. Q96L50
Gene Alias 2410005L11Rik; 4-1BBlrr; 4-1BB-mediated signaling molecule; 4-1BB-mediated-signaling molecule; AU021780; cyclophilin-like 5; leucine rich repeat protein 1; leucine-rich repeat protein 1; LRR1; LRR-1; LRR-repeat protein 1; peptidylprolyl isomerase (cyclophilin) like 5; peptidylprolyl isomerase (cyclophilin)-like 5, isoform 3; peptidylprolyl isomerase-like 5; PPIL5
Gene Symbols LRR1
Host Species Rabbit
Immunogen Recombinant Human LRR1. Recombinant protein control fragment (Product #RP-105474).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 122769
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.