Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRC10B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561558
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561558 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561558 Supplier Invitrogen™ Supplier No. PA561558
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EIEELRELRI LALDFNKLER LPDGLCRLPR LTRLYLGGNR LLALPADFAQ LQSLRCLWIE GNFLRRF Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

LRRC10B is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC10B
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LRRC10B
Gene Accession No. A6NIK2
Gene Alias Gm705; leucine rich repeat containing 10B; leucine-rich repeat-containing protein 10B; LOW QUALITY PROTEIN: leucine-rich repeat-containing protein 10B; LRRC10B; LRRC10-like protein ENSP00000367315; RGD1564983; SerdinR
Gene Symbols LRRC10B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LRRC10B. Recombinant protein control fragment (Product #RP-97562).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 390205
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.