Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRC14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120526
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120526 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120526 Supplier Invitrogen™ Supplier No. PA5120526
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: U-87MG. Immunogen sequence: MHTLVFLSTR QVLQCQPAAC QALPLLPREL FPLLFKVAFM DKKTVVLREL VHTWPFPLLS FQQLLQECAH CSRALLQERP STESMQAVIL GLTARLHTSE PGASTQPLCR KHALRVLDMT GLLDDGVEQD PGTMSMWDCT AAVARTCIAQ QQGGAAEPGP APIPVEVRVD LRVNRASYAF LREALRSSVG SPLRLCCRDL RAEDLPMRNT VALLQLLDAG CLRRVDLRFN NLGLRGLSVI IPHVARFQHL ASLRLHYVHG DSRQPSVDGE DNFRYFLAQM.

TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC14
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.26 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene LRRC14
Gene Accession No. Q15048
Gene Alias KIAA0014; leucine rich repeat containing 14; leucine-rich repeat-containing protein 14; LRRC14; LRRC14A
Gene Symbols LRRC14
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human LRRC14 (NP_055480.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9684
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.