Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRC14B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558227
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558227 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558227 Supplier Invitrogen™ Supplier No. PA558227
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LRCIEFPVPK DCYPEGAAYP QDELAMSKFN QQKYDEIAEE LRAVLLRADR EDIQVSTPLF GSFDPDIQET SNELGAFLLQ AFKTAL Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 86%.

The protein encoded by this gene is a leucine-rich repeat containing protein that is a member of the PRAME family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC14B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LRRC14B
Gene Accession No. A6NHZ5
Gene Alias AI595366; leucine rich repeat containing 14B; leucine-rich repeat-containing protein 14B; LRRC14B; RGD1564978
Gene Symbols LRRC14B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LRRC14B. Recombinant protein control fragment (Product #RP-96976).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 389257
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.