Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRRC15 Antibody (2944A), DyLight 488, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB11324K
Description
LRRC15 Monoclonal antibody specifically detects LRRC15 in Human samples. It is validated for Western Blot, Western Blot, Flow CytometrySpecifications
LRRC15 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
131578 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Flow Cytometry | |
2944A | |
Western Blot, Simple Western, Flow Cytometry | |
hLib, leucine rich repeat containing 15 | |
Chinese Hamster Ovary cell line, CHO-derived human LRRC15, Met1-Gly538, Accession # Q8TF66 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction