Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRC16A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556044
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556044 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556044 Supplier Invitrogen™ Supplier No. PA556044
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HKLADHFSRR GKTLPQQESL EIELAEEKPV KRSIITVEEL TEIERLEDLD TCMMTPKSKR KSIHSRMLRP VSRAFEMEFD L Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 83%.

Cell membrane-cytoskeleton-associated protein that plays a role in the regulation of actin polymerization at the barbed end of actin filaments. Prevents F-actin heterodimeric capping protein (CP) activity at the leading edges of migrating cells, and hence generates uncapped barbed ends and enhances actin polymerization, however, seems unable to nucleate filaments (PubMed:16054028). Plays a role in lamellipodial protrusion formations and cell migration (PubMed:19846667). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC16A
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CARMIL1
Gene Accession No. Q5VZK9
Gene Alias 1110037D04Rik; AI425970; capping protein regulator and myosin 1 linker 1; Capping protein regulator and myosin 1 linker protein 1; Capping protein, Arp2/3 and myosin-I linker homolog 1; capping protein, Arp2/3 and myosin-I linker protein 1; capping protein, Arp2/3, and Myosin-I Linker homolog 1; CARMIL; CARMIL homolog; CARMIL1; CARMIL1a; CARML1; D130057M20; dJ501N12.1; dJ501N12.5; F-actin-uncapping protein LRRC16A; leucine rich repeat containing 16; leucine rich repeat containing 16A; leucine-rich repeat-containing protein 16A; Lrrc16; LRRC16A; testicular tissue protein Li 107
Gene Symbols CARMIL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LRRC16A. Recombinant protein control fragment (Product #RP-94631).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55604
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.