Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LRRC16A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556045
Description
Immunogen sequence: RSDSKSSPQA GRRYGVQVMG SGLLAEMKAK QEKRAACAQK KLGNDAVSQD SSSPALSSVE RSDGGGAVPK LHPGLPENRF GLGTPEKNTK AEPKAEA Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 77%.
Cell membrane-cytoskeleton-associated protein that plays a role in the regulation of actin polymerization at the barbed end of actin filaments. Prevents F-actin heterodimeric capping protein (CP) activity at the leading edges of migrating cells, and hence generates uncapped barbed ends and enhances actin polymerization, however, seems unable to nucleate filaments (PubMed:16054028). Plays a role in lamellipodial protrusion formations and cell migration (PubMed:19846667). [UniProt].
Specifications
LRRC16A | |
Polyclonal | |
Unconjugated | |
CARMIL1 | |
1110037D04Rik; AI425970; capping protein regulator and myosin 1 linker 1; Capping protein regulator and myosin 1 linker protein 1; Capping protein, Arp2/3 and myosin-I linker homolog 1; capping protein, Arp2/3 and myosin-I linker protein 1; capping protein, Arp2/3, and Myosin-I Linker homolog 1; CARMIL; CARMIL homolog; CARMIL1; CARMIL1a; CARML1; D130057M20; dJ501N12.1; dJ501N12.5; F-actin-uncapping protein LRRC16A; leucine rich repeat containing 16; leucine rich repeat containing 16A; leucine-rich repeat-containing protein 16A; Lrrc16; LRRC16A; testicular tissue protein Li 107 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55604 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.90 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5VZK9 | |
CARMIL1 | |
Recombinant protein corresponding to Human LRRC16A. Recombinant protein control fragment (Product #RP-94632). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction