Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRRC16A Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548979
Description
Binds CAPZA2 with high affinity and significantly decreases CAPZA2 affinity for actin barbed ends. Increases the rate of elongation from seeds in the presence of CAPZA2, however, seems unable to nucleate filaments. Rapidly uncaps barbed ends capped by CAPZA2 and enhances barbed-end actin polymerization. May control actin dynamics in lamellipodia. Required for cell migration.
Specifications
LRRC16A | |
Polyclonal | |
Unconjugated | |
CARMIL1 | |
Leucine-rich repeat-containing protein 16A, CARMIL homolog, LRRC16A, CARMIL, CARMIL1a, LRRC16 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55604, 68732 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q5VZK9, Q6EDY6 | |
CARMIL1 | |
KLH conjugated synthetic peptide between 1152-1178 amino acids from the C-terminal region of human LRRC16A. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction