Learn More
Invitrogen™ LRRC16B Polyclonal Antibody

Description
LRRC16B Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | LRRC16B |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | CARMIL3 |
| Gene Accession No. | Q3UFQ8, Q8ND23 |
| Gene Alias | C14orf121; capping protein regulator and myosin 1 linker 3; capping protein regulator and myosin 1 linker protein 3; Capping protein, Arp2/3 and myosin-I linker protein 3; CARMIL3; crml-1; leucine rich repeat containing 16B; leucine-rich repeat-containing protein 16B; LRRC16B; mFLJ00240 |
| Gene Symbols | CARMIL3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.