Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRC16B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556518
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556518 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556518 Supplier Invitrogen™ Supplier No. PA556518
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CDYNGLHCRE EVQWDVDTIY HAEDNREFNL LDFSHLESRD LALMVAALAY NQWFTKLYCK DLRLGSEVLE QVLHTLSKSG SLEELVLDNA GLKTDFVQKL AGVFGENGSC VLHALTL Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 100%.

CARMIL3 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC16B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CARMIL3
Gene Accession No. Q8ND23
Gene Alias C14orf121; capping protein regulator and myosin 1 linker 3; capping protein regulator and myosin 1 linker protein 3; Capping protein, Arp2/3 and myosin-I linker protein 3; CARMIL3; crml-1; leucine rich repeat containing 16B; leucine-rich repeat-containing protein 16B; LRRC16B; mFLJ00240
Gene Symbols CARMIL3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LRRC16B. Recombinant protein control fragment (Product #RP-89111).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90668
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.