Learn More
Invitrogen™ LRRC17 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113953
Description
Immunogen sequence: YVFPIQTLDC KRKELKKVPN NIPPDIVKLD LSYNKINQLR PKEFEDVHEL For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 100%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 86%; Rabbit: 93%; Rat: 100%; Zebrafish: 79%.
Leucine-rich repeat-containing protein 17 (LRRC17) is an LRR protein predominantly expressed by osteoblasts. LRRC17 acts as a negative regulator of receptor activator of NF-kappaB ligand (RANKL)-induced osteoclast differentiation. It has been reported that low plasma level of LRRC17 is an independent risk factor for osteoporotic fractures.
Specifications
LRRC17 | |
Polyclonal | |
Unconjugated | |
LRRC17 | |
37 kDa leucine-rich repeat (LRR) protein; 37kDa; 4833425M04Rik; 6130400C22Rik; leucine rich repeat containing 17; leucine-rich repeat (LRR) protein; leucine-rich repeat-containing protein 17; Lrrc17; p37NB; UNQ3076/PRO9909 | |
Rabbit | |
Affinity chromatography | |
RUO | |
10234 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8N6Y2 | |
LRRC17 | |
Synthetic peptide directed towards the middle region of human LRRC17. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.