Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LRRC18 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5145142
Description
Immunogen sequence: PVELKQLKNI RAVNLGLNHL DSVPTTLGAL KELHEVGLHD NLLNNIPVSI SKLPKLKKLN IKRNPFPKPG ES Antigen sequence identity: Mouse 88%; Rat 89%.
May be involved in the regulation of spermatogenesis and sperm maturation.
Specifications
LRRC18 | |
Polyclonal | |
Unconjugated | |
Lrrc18 | |
4930442L21Rik; leucine rich repeat containing 18; leucine-rich repeat-containing protein 18; LRRC18; Mtlr1; testis-specific LRR protein; UNQ933; UNQ9338; UNQ9338/PRO34010; VKGE9338 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
474354 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N456 | |
Lrrc18 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 114-185. Recombinant protein control fragment (Product #RP-109840). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction