Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559066
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559066 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559066 Supplier Invitrogen™ Supplier No. PA559066
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LREWYISNTL IQIIPTYIQL FQAMRILDLP KNQISHLPAE IGCLKNLKEL NVGFNYLKSI PPELGDCENL ERLDCSGN Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 83%.

This gene encodes a member of the leucine-rich repeat-containing family of proteins, which function in diverse biological pathways. This family member may possibly be a nuclear protein. Similarity to the RAS suppressor protein, as well as expression down-regulation observed in tumor cells, suggests that it may function as a tumor suppressor. The gene is located in the chromosome 3 common eliminated region 1 (C3CER1), a 1.4 Mb region that is commonly deleted in diverse tumors. A related pseudogene has been identified on chromosome 2. [provided by RefSeq, Sep 2011].
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LRRC2
Gene Accession No. Q9BYS8
Gene Alias 2400002D05Rik; 4933431K03Rik; leucine rich repeat containing 2; leucine-rich repeat-containing 2; leucine-rich repeat-containing protein 2; LRRC2
Gene Symbols LRRC2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LRRC2. Recombinant protein control fragment (Product #RP-96413).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79442
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.