Learn More
Invitrogen™ LRRC2 Polyclonal Antibody

Description
Immunogen sequence: GHKVVVFDIS VIRALWETRV KKHKAWQKKE VERLEKSALE KIKEEWNFVA For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 93%; Guinea Pig: 92%; Horse: 93%; Human: 100%; Mouse: 93%; Pig: 93%; Rabbit: 93%; Rat: 93%.
Specifications
Specifications
| Antigen | LRRC2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | LRRC2 |
| Gene Accession No. | Q9BYS8 |
| Gene Alias | 2400002D05Rik; 4933431K03Rik; leucine rich repeat containing 2; leucine-rich repeat-containing 2; leucine-rich repeat-containing protein 2; LRRC2 |
| Gene Symbols | LRRC2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.