Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRRC24 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | LRRC24 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
LRRC24 Polyclonal specifically detects LRRC24 in Human samples. It is validated for Western Blot.Specifications
LRRC24 | |
Polyclonal | |
Rabbit | |
leucine rich repeat containing 24, leucine-rich repeat-containing protein 24, MGC111484 | |
LRRC24 | |
IgG |
Western Blot | |
Unconjugated | |
RUO | |
441381 | |
Synthetic peptides corresponding to LRRC24(leucine rich repeat containing 24) The peptide sequence was selected from the N terminal of LRRC24. Peptide sequence HLPRLQELHLQENSIELLEDQALAGLSSLALLDLSRNQLGTISREALQPL. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title