Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRRC25 Antibody (OTI1A2), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272529JF549
Description
LRRC25 Monoclonal specifically detects LRRC25 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
LRRC25 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
LRRC25 | |
Full length recombinant protein of human LRRC25 (NP_660299) produced in HEK293T cells. | |
0.1 mL | |
126364.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI1A2 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
leucine rich repeat containing 25, leucine-rich repeat-containing protein 25, MAPAFLJ38116, monocyte and plasmacytoid activated molecule, Monocyte and plasmacytoid-activated protein | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction