Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRC25 Monoclonal Antibody (OTI2A3), TrueMAB™, OriGene
SDP

Catalog No. 502540045
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0045 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-0045 Supplier OriGene Supplier No. TA504902S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LRRC25 Monoclonal antibody specifically detects LRRC25 in Canine, Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen LRRC25
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2A3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene COMMD1
Gene Accession No. Q8K4M5, Q8N668, Q8WMD0
Gene Alias leucine rich repeat containing 25; leucine-rich repeat-containing protein 25; LRRC25; MAPA; monocyte and plasmacytoid activated molecule; monocyte and plasmacytoid-activated protein; UNQ6169/PRO20174
Host Species Mouse
Immunogen Full length human recombinant protein of human COMMD1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150684, 17846, 289831, 403590
Target Species Canine, Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.