Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRC25 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5106995

Catalog No. PIPA5106995


Only null left
Add to Cart

Description

Description

LRRC25 Polyclonal Antibody for Western Blot

Plays a role in the inhibition of RLR-mediated type I interferon signaling pathway by targeting DDX58/RIG-I for autophagic degradation. Interacts specifically with ISG15-associated DDX58 to promote interaction between DDX58 and the autophagic cargo receptor p62/SQSTM1 to mediate DDX58 degradation via selective autophagy (PubMed:29288164). Plays also a role in the inhibition of NF-kappa-B signaling pathway and inflammatory response by promoting the degradation of p65/RELA. [UniProt].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LRRC25
Polyclonal
Unconjugated
LRRC25
leucine rich repeat containing 25; leucine-rich repeat-containing protein 25; LRRC25; MAPA; monocyte and plasmacytoid activated molecule; monocyte and plasmacytoid-activated protein; UNQ6169/PRO20174
Rabbit
Affinity Chromatography
RUO
126364
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Q8N386
LRRC25
Synthetic peptide directed towards the N terminal region of human LRRC25.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.