Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRC25 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982155
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2155 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2155 Supplier Bioss Supplier No. BS12339R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May be involved in the activation of cells of innate and acquired immunity.

Specifications

Antigen LRRC25
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene LRRC25
Gene Accession No. Q8N386
Gene Alias leucine rich repeat containing 25; Leucine-rich repeat-containing protein 25; LRRC25; MAPA; monocyte and plasmacytoid activated molecule; Monocyte and plasmacytoid-activated protein; UNQ6169/PRO20174
Gene Symbols LRRC25
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human LRRC25.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126364
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.