Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRC26 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA538058
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA538058 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA538058 Supplier Invitrogen™ Supplier No. PA538058
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A recommended positive control is human prostate tissue lysate.

The LRRC26 protein was initially identified as a cytokeratin-associated protein whose expression was elevated in multiple cancer cell lines and cancer specimens as well in normal prostate and salivary gland tissues. LRRC26 was later found to negatively regulate the NF-kappaB activation and suppresses tumor growth and metastasis. LRRC26 can also act to selectively alter the efficacy of large conductance calcium-activated K (BK) channels.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC26
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene LRRC26
Gene Accession No. Q2I0M4
Gene Alias bA350O14.10; BK channel auxiliary gamma subunit LRRC26; BK channel auxilliary gamma subunit LRRC26; CAPC; cytokeratin associated protein; cytokeratin-associated protein in cancer; leucine rich repeat containing 26; leucine-rich repeat-containing protein 26; Lrrc26; RGD1308398
Gene Symbols LRRC26
Host Species Rabbit
Immunogen 18 amino acid peptide near the carboxy terminus of human LRRC26.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 389816
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.