Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LRRC28 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566824
Description
Immunogen sequence: KRNSLTSLPEN LAQKLPNLVE LYLHSNNIVV VPEAIGSLVK LQCLDLSDNA LEIVCPEIGR LRALRHLRLA NNQLQF Highest antigen sequence identity to the following orthologs - mouse 97%, rat 99%.
LRRC28 contains 11 LRR (leucine-rich) repeats. The function of the LRRC28 protein is not known.
Specifications
LRRC28 | |
Polyclonal | |
Unconjugated | |
LRRC28 | |
1300004K21Rik; 2210012C09Rik; 2310058O11Rik; leucine rich repeat containing 28; leucine-rich repeat-containing protein 28; LRRC28; UNQ436/PRO867 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
123355 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86X40 | |
LRRC28 | |
Recombinant Human LRRC28. Recombinant protein control fragment (Product #RP-107482). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction